The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177591.1 | NP_001171062.1 | 9 - 10 | 1121 | 169 |
NM_002717.3 | NP_002708.1 | 9 - 10 | 1389 | 169 | |
XM_005273559.1 | XP_005273616.1 | 9 - 10 | 1354 | 169 | |
XM_005273560.1 | XP_005273617.1 | 10 - 11 | 1437 | 169 | |
XM_011544565.1 | XP_011542867.1 | 9 - 10 | 1745 | 169 | |
XM_011544566.1 | XP_011542868.1 | 9 - 10 | 1376 | 169 | |
XM_011544567.2 | XP_011542869.1 | 10 - 11 | 1428 | 169 | |
XM_011544568.1 | XP_011542870.1 | 9 - 10 | 1365 | 169 | |
XM_011544569.1 | XP_011542871.1 | 9 - 10 | 1097 | 169 | |
GenBank mRNA | AK095153.1 | - | 3 - 4 | 1357 | 169 |
AK303981.1 | - | 9 - 10 | 1123 | 169 | |
AK314823.1 | - | 9 - 10 | 1352 | 169 | |
AY189686.1 | - | 9 - 10 | 1086 | 169 | |
BC041071.1 | - | 9 - 10 | 1339 | 169 | |
M64929.1 | - | 9 - 10 | 1165 | 169 |