The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021934.4 | NP_068753.2 | 1 - 2 | 268 | 80 |
GenBank mRNA | AF218031.1 | - | 1 - 2 | 209 | 80 |
AK021835.1 | - | 1 - 2 | 266 | 80 | |
BC005151.2 | - | 1 - 2 | 220 | 80 | |
BC009937.2 | - | 1 - 2 | 202 | 80 | |
CB137838.1 | - | 1 - 2 | 268 | 80 |