Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCEA3
Entrez Gene ID
6920
Gene Name
transcription elongation factor A3
Gene Aliases
TFIIS, TFIIS.H
Chromosome Location
Chr.1: 23379953 - 23424779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003196.2 NP_003187.1 5 - 6 594 71
XM_006710864.1 XP_006710927.1 5 - 6 567 71
XM_011542053.2 XP_011540355.1 5 - 6 567 71
XM_017002200.1 XP_016857689.1 5 - 6 567 71
XM_017002201.1 XP_016857690.1 4 - 5 528 71
XM_017002203.1 XP_016857692.1 5 - 6 654 71
GenBank mRNA AJ223473.1 - 4 - 5 364 71
AK027024.1 - 5 - 6 555 71
AK290272.1 - 5 - 6 591 71
AK310039.1 - 4 - 5 528 71
BC041613.1 - 5 - 6 477 71
BQ949868.1 - 5 - 6 606 71

View Full Product Details