The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006946.2 | NP_008877.1 | 12 - 13 | 1728 | 56 |
XM_005274192.4 | XP_005274249.1 | 13 - 14 | 2390 | 56 | |
XM_005274193.3 | XP_005274250.1 | 12 - 13 | 2164 | 56 | |
XM_006718669.3 | XP_006718732.1 | 11 - 12 | 4077 | 56 | |
XM_006718671.3 | XP_006718734.1 | 13 - 14 | 1790 | 56 | |
XM_011545216.2 | XP_011543518.1 | 11 - 12 | 4087 | 56 | |
XM_011545217.2 | XP_011543519.1 | 11 - 12 | 4087 | 56 | |
XM_017018174.1 | XP_016873663.1 | 13 - 14 | 2482 | 56 | |
XM_017018175.1 | XP_016873664.1 | 13 - 14 | 2111 | 56 | |
XM_017018176.1 | XP_016873665.1 | 13 - 14 | 1831 | 56 | |
XM_017018177.1 | XP_016873666.1 | 12 - 13 | 2020 | 56 | |
XM_017018178.1 | XP_016873667.1 | 12 - 13 | 1703 | 56 | |
GenBank mRNA | AB008567.1 | - | 12 - 13 | 1728 | 56 |
BC139788.1 | - | 12 - 13 | 1699 | 56 |