Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPTBN2
Entrez Gene ID
6712
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
GTRAP41, SCA5, SCAR14
Chromosome Location
Chr.11: 66682496 - 66729361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006946.2 NP_008877.1 24 - 25 5056 64
XM_005274192.4 XP_005274249.1 25 - 26 5718 64
XM_005274193.3 XP_005274250.1 24 - 25 5492 64
XM_006718669.3 XP_006718732.1 23 - 24 7405 64
XM_006718671.3 XP_006718734.1 25 - 26 5118 64
XM_017018174.1 XP_016873663.1 25 - 26 5810 64
XM_017018175.1 XP_016873664.1 25 - 26 5439 64
XM_017018176.1 XP_016873665.1 25 - 26 5159 64
XM_017018177.1 XP_016873666.1 24 - 25 5348 64
XM_017018178.1 XP_016873667.1 24 - 25 5031 64
GenBank mRNA AB008567.1 - 24 - 25 5056 64
BC131819.1 - 8 - 9 1318 64

View Full Product Details