Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LSMEM1
Entrez Gene ID
286006
Gene Name
leucine rich single-pass membrane protein 1
Gene Aliases
C7orf53
Chromosome Location
Chr.7: 112480219 - 112491062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC031976.1 - 3 - 4 441 153

View Full Product Details