Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CSNK1G3
- Entrez Gene ID
-
1456
- Gene Name
- casein kinase 1 gamma 3
- Gene Aliases
-
CKI-gamma 3, CSNK1G3L
- Chromosome Location
-
Chr.5: 123512099 - 123617045 on Build GRCh38
View Full Product Details