The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195144.1 | NP_001182073.1 | 14 - 15 | 1641 | 88 |
XM_005246945.1 | XP_005247002.1 | 14 - 15 | 1685 | 88 | |
XM_005246946.1 | XP_005247003.1 | 14 - 15 | 1631 | 88 | |
XM_005246947.2 | XP_005247004.1 | 14 - 15 | 1685 | 88 | |
XM_005246948.2 | XP_005247005.1 | 14 - 15 | 1685 | 88 | |
XM_006712838.3 | XP_006712901.1 | 14 - 15 | 1685 | 88 | |
XM_011512148.2 | XP_011510450.1 | 14 - 15 | 1685 | 88 | |
XM_017005252.1 | XP_016860741.1 | 14 - 15 | 1631 | 88 | |
GenBank mRNA | AK097086.1 | - | 14 - 15 | 1450 | 88 |
AK301209.1 | - | 13 - 14 | 1562 | 88 | |
AL133087.1 | - | 8 - 9 | 894 | 88 | |
BC016985.2 | - | 14 - 15 | 1615 | 88 |