The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004398.3 | NP_004389.2 | 3 - 4 | 505 | 76 |
GenBank mRNA | AB040537.1 | - | 3 - 4 | 441 | 76 |
AB209884.1 | - | 3 - 4 | 408 | 76 | |
AK315216.1 | - | 3 - 4 | 436 | 76 | |
BC049217.1 | - | 3 - 4 | 401 | 76 | |
BC091521.1 | - | 3 - 4 | 453 | 76 | |
BC093654.1 | - | 3 - 4 | 409 | 76 | |
BC093656.1 | - | 3 - 4 | 409 | 76 | |
U28042.1 | - | 3 - 4 | 441 | 76 |