The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006078.3 | NP_006069.1 | 1 - 2 | 282 | 112 |
GenBank mRNA | AF096322.1 | - | 1 - 2 | 213 | 112 |
BC069612.1 | - | 1 - 2 | 282 | 112 | |
BC112297.1 | - | 1 - 2 | 273 | 112 | |
BC112299.1 | - | 1 - 2 | 273 | 112 | |
CR456414.1 | - | 1 - 2 | 248 | 112 |