The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114397.1 | NP_001107869.1 | 20 - 21 | 2411 | 97 |
NM_020865.2 | NP_065916.2 | 20 - 21 | 2453 | 97 | |
GenBank mRNA | AB040921.1 | - | 18 - 19 | 1905 | 97 |
AF217190.1 | - | 20 - 21 | 2445 | 97 | |
AJ577133.1 | - | 20 - 21 | 2372 | 97 | |
AJ577134.1 | - | 20 - 21 | 2330 | 97 | |
AK303720.1 | - | - | 2333 | 97 | |
AK314435.1 | - | 20 - 21 | 2453 | 97 | |
BC036035.1 | - | 19 - 20 | 2330 | 97 |