Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 57861243 - 57955323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271875.1 NP_001258804.1 4 - 5 1005 112
NM_001292025.1 NP_001278954.1 4 - 5 1310 112
XM_006721900.2 XP_006721963.1 4 - 5 1620 112
XM_011524812.2 XP_011523114.1 4 - 5 393 112
XM_017024640.1 XP_016880129.1 4 - 5 975 112
XM_017024641.1 XP_016880130.1 5 - 6 1022 112
GenBank mRNA AK000746.1 - 4 - 5 1310 112
AK000977.1 - 3 - 4 744 112
BC056882.1 - 4 - 5 1005 112

View Full Product Details