Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUEDC1
Entrez Gene ID
404093
Gene Name
CUE domain containing 1
Gene Aliases
-
Chromosome Location
Chr.17: 57861243 - 57955323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271875.1 NP_001258804.1 5 - 6 1198 81
NM_001292025.1 NP_001278954.1 5 - 6 1503 81
XM_006721900.2 XP_006721963.1 5 - 6 1813 81
XM_011524812.2 XP_011523114.1 5 - 6 586 81
XM_017024641.1 XP_016880130.1 6 - 7 1215 81
GenBank mRNA AK000746.1 - 5 - 6 1503 81
BC056882.1 - 5 - 6 1198 81

View Full Product Details