The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021110.3 | NP_066933.1 | 32 - 33 | 4236 | 110 |
XM_005251059.3 | XP_005251116.1 | 32 - 33 | 4236 | 110 | |
XM_006716651.3 | XP_006716714.1 | 32 - 33 | 4099 | 110 | |
XM_017013809.1 | XP_016869298.1 | 32 - 33 | 4025 | 110 | |
GenBank mRNA | BC014640.1 | - | 11 - 12 | 1235 | 110 |
BC140893.1 | - | 31 - 32 | 4005 | 110 | |
Y11710.1 | - | 7 - 8 | 893 | 110 |