The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021110.3 | NP_066933.1 | 25 - 26 | 3334 | 88 |
XM_005251059.3 | XP_005251116.1 | 25 - 26 | 3334 | 88 | |
XM_006716651.3 | XP_006716714.1 | 25 - 26 | 3197 | 88 | |
XM_017013809.1 | XP_016869298.1 | 25 - 26 | 3123 | 88 | |
GenBank mRNA | AK293080.1 | - | 25 - 26 | 3304 | 88 |
AK307716.1 | - | 25 - 26 | 3298 | 88 | |
AK311498.1 | - | 25 - 26 | 3298 | 88 | |
BC014640.1 | - | 4 - 5 | 332 | 88 | |
BC140893.1 | - | 24 - 25 | 3103 | 88 |