Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location
Chr.9: 137618988 - 137836127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145527.1 NP_001138999.1 10 - 11 1687 68
NM_024757.4 NP_079033.4 10 - 11 1687 68
XM_005266105.4 XP_005266162.1 10 - 11 1730 68
XM_005266110.1 XP_005266167.1 9 - 10 1602 68
XM_006717288.2 XP_006717351.1 10 - 11 1648 68
XM_011519021.2 XP_011517323.1 10 - 11 2685 68
XM_011519022.2 XP_011517324.1 10 - 11 2140 68
XM_011519023.2 XP_011517325.1 10 - 11 2664 68
XM_011519024.2 XP_011517326.1 9 - 10 2608 68
XM_011519025.1 XP_011517327.1 10 - 11 1850 68
XM_011519027.2 XP_011517329.1 10 - 11 2686 68
XM_011519028.2 XP_011517330.1 10 - 11 2686 68
XM_011519029.2 XP_011517331.1 2 - 3 275 68
XM_017015134.1 XP_016870623.1 10 - 11 2118 68
XM_017015135.1 XP_016870624.1 10 - 11 1670 68
XM_017015136.1 XP_016870625.1 9 - 10 1592 68
XM_017015137.1 XP_016870626.1 10 - 11 1829 68
XM_017015138.1 XP_016870627.1 9 - 10 1581 68
GenBank mRNA AB028932.1 - 5 - 6 507 68
AB058779.2 - 5 - 6 507 68
AK022941.1 - 9 - 10 1602 68
AY083210.1 - 8 - 9 1557 68

View Full Product Details