Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location
Chr.9: 137618988 - 137836127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145527.1 NP_001138999.1 14 - 15 2310 108
NM_024757.4 NP_079033.4 14 - 15 2310 108
XM_005266105.4 XP_005266162.1 14 - 15 2353 108
XM_005266110.1 XP_005266167.1 13 - 14 2225 108
XM_006717288.2 XP_006717351.1 14 - 15 2271 108
XM_011519021.2 XP_011517323.1 14 - 15 3308 108
XM_011519022.2 XP_011517324.1 14 - 15 2763 108
XM_011519023.2 XP_011517325.1 14 - 15 3287 108
XM_011519024.2 XP_011517326.1 13 - 14 3231 108
XM_011519025.1 XP_011517327.1 14 - 15 2473 108
XM_011519026.2 XP_011517328.1 13 - 14 3166 108
XM_011519027.2 XP_011517329.1 14 - 15 3309 108
XM_011519028.2 XP_011517330.1 14 - 15 3309 108
XM_011519029.2 XP_011517331.1 6 - 7 898 108
XM_011519033.2 XP_011517335.1 13 - 14 3145 108
XM_017015134.1 XP_016870623.1 14 - 15 2741 108
XM_017015135.1 XP_016870624.1 14 - 15 2293 108
XM_017015136.1 XP_016870625.1 13 - 14 2215 108
XM_017015137.1 XP_016870626.1 14 - 15 2452 108
XM_017015138.1 XP_016870627.1 13 - 14 2204 108
GenBank mRNA AB028932.1 - 9 - 10 1130 108
AB058779.2 - 9 - 10 1130 108
AK022941.1 - 13 - 14 2225 108
AY083210.1 - 12 - 13 2180 108
BC011608.2 - 4 - 5 527 108
BC047504.1 - 14 - 15 2310 108
BM681619.1 - 2 - 3 415 108

View Full Product Details