Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EHMT1
Entrez Gene ID
79813
Gene Name
euchromatic histone lysine methyltransferase 1
Gene Aliases
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location
Chr.9: 137618988 - 137836127 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145527.1 NP_001138999.1 9 - 10 1543 70
NM_024757.4 NP_079033.4 9 - 10 1543 70
XM_005266105.4 XP_005266162.1 9 - 10 1586 70
XM_005266110.1 XP_005266167.1 8 - 9 1458 70
XM_006717288.2 XP_006717351.1 9 - 10 1504 70
XM_011519021.2 XP_011517323.1 9 - 10 2541 70
XM_011519022.2 XP_011517324.1 9 - 10 1996 70
XM_011519023.2 XP_011517325.1 9 - 10 2520 70
XM_011519024.2 XP_011517326.1 8 - 9 2464 70
XM_011519025.1 XP_011517327.1 9 - 10 1706 70
XM_011519026.2 XP_011517328.1 9 - 10 2543 70
XM_011519027.2 XP_011517329.1 9 - 10 2542 70
XM_011519028.2 XP_011517330.1 9 - 10 2542 70
XM_011519033.2 XP_011517335.1 9 - 10 2522 70
XM_017015134.1 XP_016870623.1 9 - 10 1974 70
XM_017015135.1 XP_016870624.1 9 - 10 1526 70
XM_017015136.1 XP_016870625.1 8 - 9 1448 70
XM_017015137.1 XP_016870626.1 9 - 10 1685 70
XM_017015138.1 XP_016870627.1 8 - 9 1437 70
GenBank mRNA AB028932.1 - 4 - 5 363 70
AB058779.2 - 4 - 5 363 70
AK022941.1 - 8 - 9 1458 70
AY083210.1 - 7 - 8 1413 70
BC047504.1 - 9 - 10 1543 70

View Full Product Details