The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020405.4 | NP_065138.2 | 11 - 12 | 1400 | 72 |
GenBank mRNA | AF279144.2 | - | 11 - 12 | 1270 | 72 |
AF378753.1 | - | 11 - 12 | 1270 | 72 | |
AK093589.1 | - | 10 - 11 | 1699 | 72 | |
AK127539.1 | - | 10 - 11 | 1581 | 72 | |
AK303699.1 | - | 10 - 11 | 1087 | 72 | |
AK312999.1 | - | 11 - 12 | 1222 | 72 | |
AY704670.1 | - | 11 - 12 | 1634 | 72 | |
AY704671.1 | - | 12 - 13 | 1834 | 72 | |
AY704672.1 | - | 11 - 12 | 1580 | 72 | |
BC036059.1 | - | 11 - 12 | 1366 | 72 | |
KU257674.1 | - | 11 - 12 | 1128 | 72 |