The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020405.4 | NP_065138.2 | 5 - 6 | 805 | 80 |
GenBank mRNA | AF279144.2 | - | 5 - 6 | 675 | 80 |
AF378753.1 | - | 5 - 6 | 675 | 80 | |
AK093589.1 | - | 5 - 6 | 826 | 80 | |
AK127539.1 | - | 4 - 5 | 622 | 80 | |
AK303699.1 | - | 4 - 5 | 492 | 80 | |
AK312999.1 | - | 5 - 6 | 627 | 80 | |
AY704670.1 | - | 5 - 6 | 675 | 80 | |
AY704671.1 | - | 5 - 6 | 675 | 80 | |
AY704672.1 | - | 5 - 6 | 621 | 80 | |
BC036059.1 | - | 5 - 6 | 771 | 80 | |
KU257674.1 | - | 5 - 6 | 533 | 80 |