Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAL
Entrez Gene ID
161823
Gene Name
adenosine deaminase like
Gene Aliases
-
Chromosome Location
Chr.15: 43330351 - 43354555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012969.3 NP_001012987.1 4 - 5 990 144
NM_001159280.2 NP_001152752.1 5 - 6 1091 144
NM_001324364.1 NP_001311293.1 5 - 6 804 144
NM_001324365.1 NP_001311294.1 5 - 6 955 144
NM_001324366.1 NP_001311295.1 5 - 6 1091 144
NM_001324367.1 NP_001311296.1 5 - 6 1091 144
NM_001324368.1 NP_001311297.1 5 - 6 1091 144
XM_011521291.2 XP_011519593.1 4 - 5 567 144
XM_017021964.1 XP_016877453.1 5 - 6 677 144
XM_017021965.1 XP_016877454.1 5 - 6 656 144
XM_017021966.1 XP_016877455.1 4 - 5 593 144
XM_017021967.1 XP_016877456.1 3 - 4 341 144
XM_017021968.1 XP_016877457.1 5 - 6 800 144
XM_017021969.1 XP_016877458.1 4 - 5 567 144
GenBank mRNA AK298870.1 - 5 - 6 772 144
BC075857.1 - 4 - 5 619 144

View Full Product Details