The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182920.1 | NP_891550.1 | 5 - 6 | 1084 | 142 |
GenBank mRNA | AB037733.1 | - | 4 - 5 | 579 | 142 |
AF488803.1 | - | 5 - 6 | 1084 | 142 | |
AK310571.1 | - | 5 - 6 | 1395 | 142 | |
BC130578.1 | - | 5 - 6 | 1074 | 142 |