The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318781.1 | NP_001305710.1 | 6 - 7 | 1160 | 117 |
NM_182920.1 | NP_891550.1 | 7 - 8 | 1244 | 117 | |
GenBank mRNA | AB037733.1 | - | 6 - 7 | 739 | 117 |
AF261918.1 | - | 7 - 8 | 1224 | 117 | |
AF488803.1 | - | 7 - 8 | 1244 | 117 | |
AK310571.1 | - | 7 - 8 | 1555 | 117 | |
BC130578.1 | - | 7 - 8 | 1234 | 117 | |
BC171764.1 | - | 6 - 7 | 1150 | 117 |