Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS9
Entrez Gene ID
56999
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 9
Gene Aliases
-
Chromosome Location
Chr.3: 64515654 - 64688000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318781.1 NP_001305710.1 7 - 8 1265 111
NM_182920.1 NP_891550.1 8 - 9 1349 111
GenBank mRNA AB037733.1 - 7 - 8 844 111
AF261918.1 - 8 - 9 1329 111
AF488803.1 - 8 - 9 1349 111
AK310571.1 - 8 - 9 1660 111
BC130578.1 - 8 - 9 1339 111
BC171764.1 - 7 - 8 1255 111

View Full Product Details