Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.20: 62309060 - 62367318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005560.4 NP_005551.3 1 - 2 363 54
XM_006723796.3 XP_006723859.1 1 - 2 366 54
XM_006723798.3 XP_006723861.1 1 - 2 369 54
XM_011528818.2 XP_011527120.1 1 - 2 366 54
XM_011528819.2 XP_011527121.1 1 - 2 366 54
GenBank mRNA AF443072.1 - 1 - 2 363 54
BC085017.1 - 1 - 2 348 54

View Full Product Details