Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP11B
Entrez Gene ID
23200
Gene Name
ATPase phospholipid transporting 11B (putative)
Gene Aliases
ATPIF, ATPIR
Chromosome Location
Chr.3: 182793503 - 182923010 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014616.2 NP_055431.1 17 - 18 2119 169
XM_005247241.2 XP_005247298.1 17 - 18 2118 169
XM_005247243.2 XP_005247300.1 16 - 17 1790 169
XM_011512593.2 XP_011510895.1 17 - 18 2118 169
XM_011512594.2 XP_011510896.1 17 - 18 2118 169
XM_011512597.2 XP_011510899.1 15 - 16 1685 169
XM_017006008.1 XP_016861497.1 15 - 16 1682 169
XM_017006009.1 XP_016861498.1 17 - 18 1888 169
GenBank mRNA AB023173.1 - 4 - 5 349 169
AK093727.1 - 5 - 6 591 169
AK304855.1 - 7 - 8 1012 169
BC010630.1 - 3 - 4 247 169

View Full Product Details