The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014616.2 | NP_055431.1 | 7 - 8 | 909 | 89 |
XM_005247241.2 | XP_005247298.1 | 7 - 8 | 908 | 89 | |
XM_011512593.2 | XP_011510895.1 | 7 - 8 | 908 | 89 | |
XM_011512594.2 | XP_011510896.1 | 7 - 8 | 908 | 89 | |
XM_011512597.2 | XP_011510899.1 | 5 - 6 | 475 | 89 | |
XM_017006008.1 | XP_016861497.1 | 5 - 6 | 477 | 89 | |
GenBank mRNA | AF156548.1 | - | 7 - 8 | 818 | 89 |
BX489704.1 | - | 3 - 4 | 324 | 89 |