The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287031.1 | NP_001273960.1 | 3 - 4 | 378 | 95 |
NM_001287032.1 | NP_001273961.1 | 3 - 4 | 267 | 95 | |
NM_001287033.1 | NP_001273962.1 | 2 - 3 | 240 | 95 | |
NM_013442.2 | NP_038470.1 | 3 - 4 | 378 | 95 | |
GenBank mRNA | AF161458.1 | - | 3 - 4 | 252 | 95 |
AF190167.1 | - | 3 - 4 | 340 | 95 | |
AF282596.1 | - | 3 - 4 | 311 | 95 | |
AK027405.1 | - | 3 - 4 | 311 | 95 | |
AK225114.1 | - | 3 - 4 | 311 | 95 | |
AK303883.1 | - | 3 - 4 | 316 | 95 | |
AK307343.1 | - | 2 - 3 | 803 | 95 | |
BC002442.2 | - | 3 - 4 | 314 | 95 | |
BC003025.1 | - | 3 - 4 | 299 | 95 | |
BC010152.2 | - | 3 - 4 | 311 | 95 | |
BC014990.1 | - | 3 - 4 | 309 | 95 | |
BQ935988.1 | - | 3 - 4 | 255 | 95 | |
BU528995.1 | - | 2 - 3 | 137 | 95 | |
BX400542.2 | - | 3 - 4 | 265 | 95 |