The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321851.1 | NP_001308780.1 | 16 - 17 | 5633 | 147 |
NM_004239.4 | NP_004230.2 | 16 - 17 | 5636 | 147 | |
XM_017021787.1 | XP_016877276.1 | 17 - 18 | 5657 | 147 | |
XM_017021788.1 | XP_016877277.1 | 14 - 15 | 5508 | 147 | |
GenBank mRNA | AF007217.1 | - | 16 - 17 | 5610 | 147 |
AF011368.1 | - | 6 - 7 | 1701 | 147 | |
BC146845.1 | - | 16 - 17 | 5518 | 147 | |
BX641024.1 | - | 10 - 11 | 4408 | 147 | |
Y12490.1 | - | 16 - 17 | 5618 | 147 |