The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321851.1 | NP_001308780.1 | 19 - 20 | 5940 | 106 |
NM_004239.4 | NP_004230.2 | 19 - 20 | 5943 | 106 | |
XM_017021787.1 | XP_016877276.1 | 20 - 21 | 5964 | 106 | |
XM_017021788.1 | XP_016877277.1 | 17 - 18 | 5815 | 106 | |
GenBank mRNA | AF007217.1 | - | 19 - 20 | 5917 | 106 |
AF011368.1 | - | 9 - 10 | 2008 | 106 | |
BC146845.1 | - | 19 - 20 | 5825 | 106 | |
BX641024.1 | - | 13 - 14 | 4715 | 106 | |
L40380.1 | - | 3 - 4 | 319 | 106 | |
Y12490.1 | - | 19 - 20 | 5925 | 106 |