The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130852.1 | NP_001124324.1 | 5 - 6 | 935 | 58 |
NM_012116.3 | NP_036248.3 | 6 - 7 | 1073 | 58 | |
XM_005258696.3 | XP_005258753.1 | 6 - 7 | 1080 | 58 | |
XM_011526688.2 | XP_011524990.1 | 6 - 7 | 1081 | 58 | |
XM_011526689.2 | XP_011524991.1 | 5 - 6 | 945 | 58 | |
XM_011526690.2 | XP_011524992.1 | 5 - 6 | 981 | 58 | |
GenBank mRNA | AB028645.1 | - | 6 - 7 | 1051 | 58 |
AF117646.1 | - | 6 - 7 | 1073 | 58 | |
AF117647.1 | - | 5 - 6 | 935 | 58 | |
BC028915.1 | - | 6 - 7 | 1031 | 58 |