The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038707.1 | NP_001033796.1 | 3 - 4 | 621 | 57 |
NM_020239.3 | NP_064624.1 | 2 - 3 | 485 | 57 | |
XM_017001847.1 | XP_016857336.1 | 3 - 4 | 565 | 57 | |
GenBank mRNA | AF187845.2 | - | 2 - 3 | 410 | 57 |
AF286041.1 | - | 1 - 2 | 45 | 57 | |
BC012796.2 | - | 1 - 2 | 229 | 57 | |
BC041604.1 | - | 2 - 3 | 447 | 57 | |
BP340339.1 | - | 2 - 3 | 486 | 57 | |
BX339337.2 | - | 3 - 4 | 534 | 57 |