The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243254.1 | NP_001230183.1 | 13 - 14 | 2772 | 77 |
NM_015213.3 | NP_056028.2 | 13 - 14 | 2772 | 77 | |
XM_005252832.1 | XP_005252889.1 | 13 - 14 | 2781 | 77 | |
XM_011519953.2 | XP_011518255.1 | 8 - 9 | 1035 | 77 | |
GenBank mRNA | AB029014.1 | - | 13 - 14 | 2732 | 77 |
AK025499.1 | - | 8 - 9 | 1383 | 77 | |
AK026007.1 | - | 6 - 7 | 848 | 77 | |
AK125444.1 | - | 5 - 6 | 2077 | 77 | |
AK295881.1 | - | 13 - 14 | 2533 | 77 | |
AK302478.1 | - | 10 - 11 | 2233 | 77 | |
AL117448.1 | - | 13 - 14 | 2528 | 77 | |
BC009354.2 | - | 9 - 10 | 1500 | 77 | |
BC071596.1 | - | 14 - 15 | 2577 | 77 |