Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
Chromosome Location
Chr.10: 109996350 - 110135565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001121.3 NP_001112.2 12 - 13 2000 71
NM_001320591.1 NP_001307520.1 13 - 14 1921 71
NM_001320592.1 NP_001307521.1 12 - 13 1984 71
NM_001320593.1 NP_001307522.1 12 - 13 1980 71
NM_001320594.1 NP_001307523.1 13 - 14 2054 71
NM_016824.4 NP_058432.1 12 - 13 2000 71
NM_019903.4 NP_063968.1 12 - 13 1984 71
GenBank mRNA AB209810.1 - 12 - 13 1763 71
AK223270.1 - 13 - 14 1590 71
AK291488.1 - 12 - 13 1816 71
BC043143.1 - 12 - 13 1725 71
BC057285.1 - 12 - 13 1710 71
BC062559.1 - 12 - 13 1973 71
BX647403.1 - 12 - 13 1983 71
BX647442.1 - 3 - 4 1461 71
BX648016.1 - 12 - 13 1953 71
D67031.1 - 12 - 13 1790 71
EF553523.1 - 13 - 14 1922 71
U37122.1 - 12 - 13 1742 71

View Full Product Details