Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
Chromosome Location
Chr.10: 109996350 - 110135565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001121.3 NP_001112.2 13 - 14 2125 128
NM_019903.4 NP_063968.1 13 - 14 2109 128
GenBank mRNA BX647403.1 - 13 - 14 2108 128
BX648016.1 - 13 - 14 2078 128
D67031.1 - 13 - 14 1915 128
U37122.1 - 13 - 14 1867 128

View Full Product Details