The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001121.3 | NP_001112.2 | 4 - 5 | 874 | 109 |
NM_001320591.1 | NP_001307520.1 | 5 - 6 | 795 | 109 | |
NM_001320592.1 | NP_001307521.1 | 4 - 5 | 858 | 109 | |
NM_001320593.1 | NP_001307522.1 | 4 - 5 | 854 | 109 | |
NM_001320594.1 | NP_001307523.1 | 5 - 6 | 928 | 109 | |
NM_016824.4 | NP_058432.1 | 4 - 5 | 874 | 109 | |
NM_019903.4 | NP_063968.1 | 4 - 5 | 858 | 109 | |
GenBank mRNA | AB209810.1 | - | 4 - 5 | 637 | 109 |
AK223270.1 | - | 5 - 6 | 464 | 109 | |
AK291488.1 | - | 4 - 5 | 690 | 109 | |
BC043143.1 | - | 4 - 5 | 599 | 109 | |
BC057285.1 | - | 4 - 5 | 584 | 109 | |
BC062559.1 | - | 4 - 5 | 847 | 109 | |
BX647403.1 | - | 4 - 5 | 857 | 109 | |
BX648016.1 | - | 4 - 5 | 827 | 109 | |
CN428885.1 | - | 5 - 6 | 599 | 109 | |
D67031.1 | - | 4 - 5 | 664 | 109 | |
EF553523.1 | - | 5 - 6 | 796 | 109 | |
U37122.1 | - | 4 - 5 | 613 | 109 |