Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADD3
Entrez Gene ID
120
Gene Name
adducin 3
Gene Aliases
ADDL, CPSQ3
Chromosome Location
Chr.10: 109996350 - 110135565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001121.3 NP_001112.2 7 - 8 1254 74
NM_001320591.1 NP_001307520.1 8 - 9 1175 74
NM_001320592.1 NP_001307521.1 7 - 8 1238 74
NM_001320593.1 NP_001307522.1 7 - 8 1234 74
NM_001320594.1 NP_001307523.1 8 - 9 1308 74
NM_016824.4 NP_058432.1 7 - 8 1254 74
NM_019903.4 NP_063968.1 7 - 8 1238 74
GenBank mRNA AB209810.1 - 7 - 8 1017 74
AK291488.1 - 7 - 8 1070 74
BC043143.1 - 7 - 8 979 74
BC057285.1 - 7 - 8 964 74
BC062559.1 - 7 - 8 1227 74
BX648016.1 - 7 - 8 1207 74
EF553523.1 - 8 - 9 1176 74
U37122.1 - 7 - 8 993 74

View Full Product Details