The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101654.1 | NP_001095124.1 | 8 - 9 | 1770 | 64 |
XM_011525940.2 | XP_011524242.1 | 9 - 10 | 1837 | 64 | |
XM_011525941.2 | XP_011524243.1 | 7 - 8 | 1515 | 64 | |
GenBank mRNA | BC015733.1 | - | 8 - 9 | 1084 | 64 |