The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153230.2 | NP_694962.1 | 3 - 4 | 1321 | 70 |
XM_011523697.1 | XP_011521999.1 | 3 - 4 | 1397 | 70 | |
GenBank mRNA | AI125619.1 | - | 1 - 2 | 70 | |
BC034782.1 | - | 3 - 4 | 1301 | 70 | |
DB524242.1 | - | 1 - 2 | 137 | 70 |