The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193376.1 | NP_001180305.1 | 1 - 2 | 278 | 119 |
NM_198253.2 | NP_937983.2 | 1 - 2 | 278 | 119 | |
GenBank mRNA | AB085628.1 | - | 1 - 2 | 220 | 119 |
AB086379.1 | - | 1 - 2 | 220 | 119 | |
AB086950.1 | - | 1 - 2 | 220 | 119 | |
AF015950.1 | - | 1 - 2 | 275 | 119 | |
AF018167.1 | - | 1 - 2 | 276 | 119 | |
AX810036.1 | - | 1 - 2 | 275 | 119 | |
AX810378.1 | - | 1 - 2 | 275 | 119 |