The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193376.1 | NP_001180305.1 | 2 - 3 | 1632 | 79 |
NM_198253.2 | NP_937983.2 | 2 - 3 | 1632 | 79 | |
XM_017009796.1 | XP_016865285.1 | 2 - 3 | 2204 | 79 | |
GenBank mRNA | AB085628.1 | - | 2 - 3 | 1574 | 79 |
AB086379.1 | - | 2 - 3 | 1574 | 79 | |
AB086950.1 | - | 2 - 3 | 1574 | 79 | |
AF015950.1 | - | 2 - 3 | 1629 | 79 | |
AF018167.1 | - | 2 - 3 | 1630 | 79 | |
AX810036.1 | - | 2 - 3 | 1629 | 79 | |
AX810378.1 | - | 2 - 3 | 1629 | 79 |