The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193376.1 | NP_001180305.1 | 9 - 10 | 2638 | 79 |
NM_198253.2 | NP_937983.2 | 9 - 10 | 2638 | 79 | |
XM_017009796.1 | XP_016865285.1 | 9 - 10 | 3210 | 79 | |
GenBank mRNA | AB085628.1 | - | 9 - 10 | 2580 | 79 |
AB086379.1 | - | 7 - 8 | 2398 | 79 | |
AB086950.1 | - | 7 - 8 | 2362 | 79 | |
AF015950.1 | - | 9 - 10 | 2635 | 79 | |
AF018167.1 | - | 9 - 10 | 2636 | 79 | |
AX810036.1 | - | 9 - 10 | 2635 | 79 | |
AX810378.1 | - | 9 - 10 | 2635 | 79 | |
BC062321.1 | - | 5 - 6 | 1211 | 79 | |
JF896283.1 | - | 2 - 3 | 250 | 79 |