Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GDI1
Entrez Gene ID
2664
Gene Name
GDP dissociation inhibitor 1
Gene Aliases
1A, GDIL, MRX41, MRX48, OPHN2, RABGD1A, RABGDIA, XAP-4
Chromosome Location
Chr.X: 154436913 - 154443467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001493.2 NP_001484.1 8 - 9 1330 65
GenBank mRNA AB102647.1 - 8 - 9 988 65
AF400433.1 - 8 - 9 995 65
AK090746.1 - 8 - 9 1078 65
AK123405.1 - 1 - 2 1268 65
AK131466.1 - 4 - 5 2709 65
AK295306.1 - 7 - 8 1041 65
AK303248.1 - 8 - 9 902 65
AL833629.1 - 8 - 9 1084 65
BC000317.2 - 8 - 9 1030 65
BC012201.1 - 8 - 9 1077 65
BT019884.1 - 8 - 9 988 65
CR542258.1 - 8 - 9 988 65
CR542276.1 - 8 - 9 988 65
D45021.1 - 8 - 9 1018 65
KU178091.1 - 8 - 9 988 65
KU178092.1 - 6 - 7 620 65
X79353.1 - 8 - 9 1068 65

View Full Product Details