The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098627.3 | NP_001092097.2 | 5 - 6 | 698 | 76 |
NM_001098630.2 | NP_001092100.1 | 5 - 6 | 605 | 76 | |
NM_032643.4 | NP_116032.1 | 5 - 6 | 521 | 76 | |
XM_011516162.1 | XP_011514464.1 | 5 - 6 | 620 | 76 | |
XM_011516163.2 | XP_011514465.1 | 5 - 6 | 806 | 76 | |
XM_011516164.1 | XP_011514466.1 | 5 - 6 | 688 | 76 | |
GenBank mRNA | AY504946.1 | - | 5 - 6 | 530 | 76 |
AY504947.1 | - | 5 - 6 | 553 | 76 | |
AY693666.1 | - | 5 - 6 | 532 | 76 | |
AY693667.1 | - | 4 - 5 | 187 | 76 | |
BC004139.2 | - | 5 - 6 | 521 | 76 | |
BC004201.2 | - | 5 - 6 | 521 | 76 | |
DQ277633.1 | - | 5 - 6 | 521 | 76 | |
DQ277634.1 | - | 5 - 6 | 502 | 76 | |
JQ950685.1 | - | 2 - 3 | 71 | 76 | |
KP843527.1 | - | 5 - 6 | 581 | 76 |