The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002428.3 | NP_002419.1 | 8 - 9 | 2242 | 61 |
GenBank mRNA | BC006412.1 | - | 8 - 9 | 1362 | 61 |
BC036495.1 | - | 8 - 9 | 2248 | 61 | |
BC055428.1 | - | 8 - 9 | 2243 | 61 | |
BT006715.1 | - | 6 - 7 | 1142 | 61 | |
D85510.1 | - | 5 - 6 | 972 | 61 | |
D86331.1 | - | 7 - 8 | 1180 | 61 | |
Z48482.1 | - | 8 - 9 | 1505 | 61 |