The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002428.3 | NP_002419.1 | 9 - 10 | 2356 | 77 |
GenBank mRNA | BC006412.1 | - | 9 - 10 | 1476 | 77 |
BC036495.1 | - | 9 - 10 | 2362 | 77 | |
BC055428.1 | - | 9 - 10 | 2357 | 77 | |
BT006715.1 | - | 7 - 8 | 1256 | 77 | |
D85510.1 | - | 6 - 7 | 1086 | 77 | |
D86331.1 | - | 8 - 9 | 1294 | 77 | |
Z48482.1 | - | 9 - 10 | 1619 | 77 |