The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040113.1 | NP_001035202.1 | 28 - 29 | 3781 | 61 |
NM_001040114.1 | NP_001035203.1 | 28 - 29 | 3781 | 61 | |
NM_002474.2 | NP_002465.1 | 27 - 28 | 3760 | 61 | |
NM_022844.2 | NP_074035.1 | 27 - 28 | 3760 | 61 | |
XM_011522502.2 | XP_011520804.1 | 27 - 28 | 3762 | 61 | |
XM_017023250.1 | XP_016878739.1 | 28 - 29 | 3745 | 61 | |
GenBank mRNA | AB020673.2 | - | 27 - 28 | 3728 | 61 |
AY520816.1 | - | 28 - 29 | 3762 | 61 | |
AY520817.1 | - | 28 - 29 | 3762 | 61 | |
BC101677.1 | - | 27 - 28 | 3724 | 61 | |
BC104906.1 | - | 27 - 28 | 3724 | 61 | |
BC143364.1 | - | 28 - 29 | 3745 | 61 | |
D10667.1 | - | 6 - 7 | 995 | 61 | |
EU489063.1 | - | 28 - 29 | 3781 | 61 | |
X69292.1 | - | 3 - 4 | 381 | 61 |