Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2B
Entrez Gene ID
2959
Gene Name
general transcription factor IIB
Gene Aliases
TF2B, TFIIB
Chromosome Location
Chr.1: 88852638 - 88891944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001514.5 NP_001505.1 2 - 3 246 72
XM_011541299.1 XP_011539601.1 2 - 3 448 72
GenBank mRNA AB451296.1 - 2 - 3 127 72
AK289822.1 - 2 - 3 192 72
BC020597.1 - 2 - 3 192 72
BP379342.1 - 2 - 3 246 72
F07886.1 - 2 - 3 198 72
M76766.1 - 2 - 3 165 72
S44184.1 - 1 - 2 115 72

View Full Product Details