The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134382.2 | NP_001127854.1 | 9 - 10 | 2419 | 72 |
NM_014869.6 | NP_055684.3 | 9 - 10 | 2605 | 72 | |
XM_011534304.2 | XP_011532606.1 | 11 - 12 | 2656 | 72 | |
XM_011534305.2 | XP_011532607.1 | 9 - 10 | 2405 | 72 | |
XM_011534306.2 | XP_011532608.1 | 9 - 10 | 2811 | 72 | |
XM_011534307.2 | XP_011532609.1 | 10 - 11 | 2757 | 72 | |
XM_011534308.2 | XP_011532610.1 | 9 - 10 | 2600 | 72 | |
XM_011534310.2 | XP_011532612.1 | 11 - 12 | 2656 | 72 | |
XM_011534311.2 | XP_011532613.1 | 9 - 10 | 2768 | 72 | |
XM_011534312.2 | XP_011532614.1 | 9 - 10 | 2396 | 72 | |
XM_011534313.2 | XP_011532615.1 | 9 - 10 | 2615 | 72 | |
XM_011534314.2 | XP_011532616.1 | 9 - 10 | 2351 | 72 | |
XM_011534315.2 | XP_011532617.1 | 9 - 10 | 2354 | 72 | |
XM_017007581.1 | XP_016863070.1 | 10 - 11 | 2626 | 72 | |
XM_017007582.1 | XP_016863071.1 | 10 - 11 | 2611 | 72 | |
GenBank mRNA | AB018306.1 | - | 8 - 9 | 2098 | 72 |
AK294525.1 | - | 9 - 10 | 2419 | 72 | |
AY653734.1 | - | 9 - 10 | 2367 | 72 | |
BC010267.1 | - | 9 - 10 | 2359 | 72 |