The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134382.2 | NP_001127854.1 | 10 - 11 | 2519 | 68 |
NM_014869.6 | NP_055684.3 | 10 - 11 | 2705 | 68 | |
XM_011534304.2 | XP_011532606.1 | 12 - 13 | 2756 | 68 | |
XM_011534305.2 | XP_011532607.1 | 10 - 11 | 2505 | 68 | |
XM_011534306.2 | XP_011532608.1 | 10 - 11 | 2911 | 68 | |
XM_011534307.2 | XP_011532609.1 | 11 - 12 | 2857 | 68 | |
XM_011534308.2 | XP_011532610.1 | 10 - 11 | 2700 | 68 | |
XM_011534310.2 | XP_011532612.1 | 12 - 13 | 2756 | 68 | |
XM_011534311.2 | XP_011532613.1 | 10 - 11 | 2868 | 68 | |
XM_011534312.2 | XP_011532614.1 | 10 - 11 | 2496 | 68 | |
XM_011534313.2 | XP_011532615.1 | 10 - 11 | 2715 | 68 | |
XM_011534314.2 | XP_011532616.1 | 10 - 11 | 2451 | 68 | |
XM_011534315.2 | XP_011532617.1 | 10 - 11 | 2454 | 68 | |
XM_017007581.1 | XP_016863070.1 | 11 - 12 | 2726 | 68 | |
XM_017007582.1 | XP_016863071.1 | 11 - 12 | 2711 | 68 | |
GenBank mRNA | AB018306.1 | - | 9 - 10 | 2198 | 68 |
AK294525.1 | - | 10 - 11 | 2519 | 68 | |
AY653734.1 | - | 10 - 11 | 2467 | 68 | |
BC010267.1 | - | 10 - 11 | 2459 | 68 |